This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459139_g1
See other LDHD GE Assays ›
Gene Symbol:
Ldhd
Gene Name:
lactate dehydrogenase D
Gene Aliases:
4733401P21Rik, D8Bwg1320e
Chromosome Location:
Chr.8: 111626271 - 111630322 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459139_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ldhd
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_027570.3 NP_081846.3 3 - 4 334 64
GenBank mRNA AK014649.1 - 3 - 4 349 64
AK029820.1 - 348 64
AY092768.1 - 3 - 4 339 64
BC039155.1 - 3 - 4 330 4242682 64
BC055443.1 - 3 - 4 334 4235345 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0800002950.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ldhd

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

ATP biosynthetic process
oxidation-reduction process
catalytic activity
D-lactate dehydrogenase (cytochrome) activity
protein binding
D-lactate dehydrogenase activity
UDP-N-acetylmuramate dehydrogenase activity
oxidoreductase activity
oxidoreductase activity, acting on CH-OH group of donors
flavin adenine dinucleotide binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0800022797.mm.1 NM_027570.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0800022797.mm.1 NM_027570.3 Refseq
Affymetrix Mouse Clariom S Array PSR0800022797.mm.1 TC0800002950.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-55658555d6-qm6d9:80/100.66.79.83:80.
git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.41.0-Offline