This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459195_g1
See other CIC GE Assays ›
Gene Symbol:
Cic
Gene Name:
capicua homolog (Drosophila)
Gene Aliases:
1200010B10Rik, mKIAA0306
Chromosome Location:
Chr.7: 25267657 - 25294159 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459195_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cic
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF363690.1 - 5 - 6 802 70
AK147276.1 - 5 - 6 1435 70
AK147535.1 - 6 - 7 3553 70
BC058665.1 - 2 - 3 320 70

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0