This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459513_g1
See other IGSF6 GE Assays ›
Gene Symbol:
Igsf6
Gene Name:
immunoglobulin superfamily, member 6
Gene Aliases:
-
Chromosome Location:
Chr.7: 121064067 - 121074903 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459513_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_030691.1 NP_109616.1 4 - 5 607 109
GenBank mRNA AJ278575.1 - 4 - 5 607 109
AK040569.1 - 4 - 5 110
AK151417.1 - 4 - 5 596 109
AK152186.1 - 4 - 5 596 109
AK156382.1 - 4 - 5 545 109
AK163058.1 - 4 - 5 110
BC057951.1 - 4 - 5 604 5341788 109

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004207.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf6

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700035032.mm.1 NM_030691.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035032.mm.1 NM_030691.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700035032.mm.1 TC0700004207.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rhr4g:80/100.66.78.163:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0