This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459614_m1
See other TRPS1 GE Assays ›
Gene Symbol:
Trps1
Gene Name:
trichorhinophalangeal syndrome I (human)
Gene Aliases:
AI115454, AI447310, D15Ertd586e
Chromosome Location:
Chr.15: 50654757 - 50890747 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459614_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Trps1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032000.2 NP_114389.2 5 - 6 3225 90
GenBank mRNA AF346836.1 - 6 - 7 3150 90
AK142355.1 - 5 - 6 2895 90
BC037058.1 - 7 - 8 2336 4951294 90
BC049857.1 - 8 - 9 2191 4460789 90
BC083110.1 - 4 - 5 1983 90
BC099962.1 - 5 - 6 3190 30091008 90

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001491.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Trps1

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
chondrocyte differentiation
transcription, DNA-templated
regulation of transcription, DNA-templated
transmembrane receptor protein serine/threonine kinase signaling pathway
regulation of histone deacetylation
regulation of chondrocyte differentiation
negative regulation of transcription, DNA-templated
positive regulation of extrinsic apoptotic signaling pathway via death domain receptors
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
sequence-specific DNA binding
metal ion binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500011348.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011348.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011348.mm.1 TC1500001491.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1500011352.mm.1 NM_032000.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500011352.mm.1 NM_032000.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500011352.mm.1 TC1500001491.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 717706 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0