This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459628_g1
See other TNFRSF25 GE Assays ›
Gene Symbol:
Tnfrsf25
Gene Name:
tumor necrosis factor receptor superfamily, member 25
Gene Aliases:
APO-3, DDR3, DR3, LARD, TR3, TRAMP, Tnfrsf12, WSL-1, WSL-LR, Wsl
Chromosome Location:
Chr.4: 152115528 - 152120119 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459628_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Tnfrsf25
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001291010.1 NP_001277939.1 8 - 9 865 76
NM_033042.4 NP_149031.2 5 - 6 815 76
GenBank mRNA AF329969.1 - 8 - 9 869 76
AK040632.1 - 5 - 6 1484 76
BC017526.1 - 5 - 6 815 4912933 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002013.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Apoptosis Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Tnfrsf25

Panther Classification:

Molecular Function -

transmembrane signal receptor

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400016488.mm.1 NM_033042.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400016488.mm.1 TC0400002013.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400016489.mm.1 NM_033042.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400016489.mm.1 TC0400002013.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline