This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459680_g1
See other IGSF9 GE Assays ›
Gene Symbol:
Igsf9
Gene Name:
immunoglobulin superfamily, member 9
Gene Aliases:
644ETD8, Dasm1, Kiaa1355-hp, NRT1, Ncaml, mKIAA1355
Chromosome Location:
Chr.1: 172481776 - 172498877 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459680_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf9
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001145800.1 NP_001139272.1 11 - 12 1622 121
NM_033608.3 NP_291086.2 11 - 12 1688 121
GenBank mRNA AF317839.1 - 11 - 12 1688 121
AK078536.1 - 11 - 12 1616 121
AK145121.1 - 11 - 12 1597 121
BC034594.1 - 11 - 12 1604 121
BC040281.1 - 11 - 12 1648 5364073 121

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100001616.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf9

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
multicellular organism development
nervous system development
dendrite development
cell differentiation
regulation of synapse organization
protein binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100013462.mm.1 NM_001145800.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100013462.mm.1 NM_001145800.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100013462.mm.1 NM_033608.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100013462.mm.1 NM_033608.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100013462.mm.1 TC0100001616.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline