This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00459993_m1
See other PRMT2 GE Assays ›
Gene Symbol:
Prmt2
Gene Name:
protein arginine N-methyltransferase 2
Gene Aliases:
AI504737, Hrmt1l1
Chromosome Location:
Chr.10: 76207226 - 76237865 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00459993_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prmt2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AF169620.1 - 3 - 4 263 76
AK077456.1 - 3 - 4 320 76
AK140200.1 - 3 - 4 347 76
AK144412.1 - 3 - 4 318 76
AK145829.1 - 3 - 4 320 76
AK146754.1 - 3 - 4 319 76
AK146966.1 - 3 - 4 327 76
AK159186.1 - 3 - 4 326 76
BC088997.1 - 3 - 4 277 76
BC122563.1 - 2 - 3 251 40107557 76
BC125275.1 - 2 - 3 251 40107558 76
BY288156.1 - 3 - 4 355 76
BY727303.1 - 3 - 4 346 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Probe spans exons

Panther Classification:

Molecular Function -

protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-c7pcq:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0