This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461251_m1
See other CTH GE Assays ›
Gene Symbol:
Cth
Gene Name:
cystathionase (cystathionine gamma-lyase)
Gene Aliases:
0610010I13Rik, AI314617, BC019483, CSE, Cys3
Chromosome Location:
Chr.3: 157894248 - 157925081 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461251_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cth
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_145953.2 NP_666065.1 5 - 6 618 120
GenBank mRNA AK002480.1 - 5 - 6 602 120
AY083352.1 - 5 - 6 618 120
BC019483.1 - 5 - 6 604 4235178 120

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300003194.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cth

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
negative regulation of cell proliferation
cellular amino acid biosynthetic process
protein-pyridoxal-5-phosphate linkage via peptidyl-N6-pyridoxal phosphate-L-lysine
cysteine biosynthetic process
negative regulation of cell growth
negative regulation of apoptotic process
positive regulation of I-kappaB kinase/NF-kappaB signaling
protein sulfhydration
homocysteine metabolic process
positive regulation of NF-kappaB transcription factor activity
protein homotetramerization
hydrogen sulfide biosynthetic process
negative regulation of apoptotic signaling pathway
catalytic activity
cystathionine beta-lyase activity
cystathionine gamma-lyase activity
protein binding
calmodulin binding
lyase activity
pyridoxal phosphate binding
L-cystine L-cysteine-lyase (deaminating)
L-cysteine desulfhydrase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0300025034.mm.1 NM_145953.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300025034.mm.1 NM_145953.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300025034.mm.1 TC0300003194.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300025035.mm.1 NM_145953.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300025035.mm.1 NM_145953.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300025035.mm.1 TC0300003194.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300025036.mm.1 NM_145953.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300025036.mm.1 NM_145953.2 Refseq
Affymetrix Mouse Clariom S Array PSR0300025036.mm.1 TC0300003194.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-5wn4c:80/100.66.76.150:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline