This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461346_g1
See other GMDS GE Assays ›
Gene Symbol:
Gmds
Gene Name:
GDP-mannose 4, 6-dehydratase
Gene Aliases:
BC031788, C87208
Chromosome Location:
Chr.13: 31819583 - 32338725 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461346_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Gmds
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146041.2 NP_666153.1 7 - 8 836 62
GenBank mRNA BC031788.1 - 7 - 8 836 4165509 62
BC093502.1 - 7 - 8 860 4215865 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300001751.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Gmds

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

GDP-mannose metabolic process
catalytic activity
GDP-mannose 4,6-dehydratase activity
lyase activity
coenzyme binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300010899.mm.1 NM_146041.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300010899.mm.1 NM_146041.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300010899.mm.1 TC1300001751.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1300010897.mm.1 NM_146041.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300010897.mm.1 NM_146041.2 Refseq
Affymetrix Mouse Clariom S Array PSR1300010897.mm.1 TC1300001751.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0