This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461491_m1
See other CMTR2 GE Assays ›
Gene Symbol:
Cmtr2
Gene Name:
cap methyltransferase 2
Gene Aliases:
AU022703, C730036L12Rik, Ftsjd1, MTr2
Chromosome Location:
Chr.8: 110215347 - 110224489 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461491_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cmtr2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146215.4 NP_666327.2 1 - 2 126 64
GenBank mRNA AK032855.1 - 1 - 2 102 64
AK050315.1 - 1 - 2 98 64
AK158368.1 - 1 - 2 108 64
BC025546.1 - 1 - 2 108 5320939 64

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK050315.1, AK158368.1, BC025546.1, NM_146215.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cmtr2

Gene Ontology Categories:

Function(s) Process(es)

7-methylguanosine mRNA capping
mRNA processing
methylation
cap2 mRNA methylation
mRNA (nucleoside-2'-O-)-methyltransferase activity
methyltransferase activity
transferase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26161388 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26161392 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26161391 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176071 BC025546.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176071 AK050315.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176071 NM_146215.4 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176071 BC025546.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176071 AK050315.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_51_P176071 NM_146215.4 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 44536 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 884303 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 529319 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 582470 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 276370 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 163797 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P168619 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176071 BC025546.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176071 AK050315.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176071 NM_146215.4 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176071 BC025546.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176071 AK050315.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_51_P176071 NM_146215.4 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161383 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161401 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161403 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161390 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161369 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161365 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161405 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161371 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161386 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161380 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161364 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161398 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161375 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161402 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161366 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161388 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161392 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161396 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161389 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161400 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161373 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161377 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161382 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161385 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161399 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P26161379 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06638870 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P168619 - -
Whole Mouse Genome G4122A A_52_P375931 BC025546.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P375931 AK050315.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P375931 NM_146215.4 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P375931 AK032855.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176071 BC025546.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176071 AK050315.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176071 NM_146215.4 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176071 BC025546.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176071 AK050315.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P176071 NM_146215.4 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P128251 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P168619 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07024678 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P07024680 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26161392 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26161388 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P26161391 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161358 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161441 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161471 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161469 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161465 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161420 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161485 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161355 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161369 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161473 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161430 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161467 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161349 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161382 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161365 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161403 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161479 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161478 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161439 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161401 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161433 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161438 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161405 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161371 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161411 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161375 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161412 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161377 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161364 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161404 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161442 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161362 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161360 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161383 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161418 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161415 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161400 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161425 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161459 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161356 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161373 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161366 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161460 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161435 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161455 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161343 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161451 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161363 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161458 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161393 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161419 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161389 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161406 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161421 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161392 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161456 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161436 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161409 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161423 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161444 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161399 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161432 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161429 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161379 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161398 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161402 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P26161345 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161433 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06926472 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161421 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161436 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07216471 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07342943 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06717303 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161435 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161401 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161463 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161369 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161360 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161343 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161358 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161376 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161354 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161423 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161406 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161430 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161398 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07024674 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161364 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161383 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161405 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06717314 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07268413 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161349 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161412 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07268404 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161362 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161469 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161363 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161389 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06829926 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161402 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161365 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161409 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161355 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07251398 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161477 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161415 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161373 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161441 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06759551 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161399 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161400 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161366 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161445 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161396 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07268411 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161442 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161467 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161419 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161439 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161438 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P07268415 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161356 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161345 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161377 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161432 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06717315 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161371 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161404 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161418 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161425 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161388 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161346 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161429 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161462 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P26161351 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10575377 - -
Affymetrix Mouse Gene 1.0 ST Array 10575378 - -
Affymetrix Mouse Gene 1.0 ST Array 10575379 - -
Affymetrix HT MG-430 PM Array Plate 1437684_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1451273_PM_x_at - -
Affymetrix HT MG-430 PM Array Plate 1424232_PM_a_at - -
Affymetrix Mouse Gene 1.1 ST Array 10575376 - -
Mouse Genome 430 2.0 Array 1437684_PM_at - -
Mouse Genome 430 2.0 Array 1424232_a_at - -
Mouse Genome 430 2.0 Array 1451273_PM_x_at - -
Mouse Genome 430 2.0 Array 1451273_x_at - -
Mouse Genome 430 2.0 Array 1424232_PM_a_at - -
Mouse Genome 430 2.0 Array 1437684_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 234728_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 234728_at - -
Mouse Expression Set 430 1437684_PM_at - -
Mouse Expression Set 430 1437684_at - -
Murine Genome U74v2 Set 103497_at - -
Murine Genome U74v2 Set 103497_PM_at - -
Mouse Genome 430A 2.0 Array 1437684_at - -
Mouse Genome 430A 2.0 Array 1437684_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1437684_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1451273_x_at - -
Affymetrix HT Mouse Genome 430A Array 1424232_PM_a_at - -
Affymetrix HT Mouse Genome 430A Array 1437684_at - -
Affymetrix HT Mouse Genome 430A Array 1451273_PM_x_at - -
Affymetrix HT Mouse Genome 430A Array 1424232_a_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 673057 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 6860086 - -
Illumina mouse-6 v1.0 expression beadchip 6860086 - -
Illumina mouseRef-8 v1.0 expression beadchip 0006860086 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2720764 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2720764 - -
mouse-6 v1.1 (Illumina) scl33327.2_154-S - -
Illumina Sentrix mouse-6.v1 bead array 6860086 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl33327.2_154-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 6860086 - -
MouseRef-8 v2 Expression BeadChip ILMN_2720763 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2720763 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl33327.2_154-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0