This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00461516_m1
See other MMGT1 GE Assays ›
Gene Symbol:
Mmgt1
Gene Name:
membrane magnesium transporter 1
Gene Aliases:
9630048L06Rik, BC032271, Emc5, Mir5116, Tmem32, mir-5116
Chromosome Location:
Chr.X: 56585512 - 56597919 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00461516_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mmgt1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_146234.3 NP_666346.1 2 - 3 187 128
GenBank mRNA AK036253.1 - 2 - 3 188 128
AK043524.1 - 2 - 3 201 128
AK049689.1 - 2 - 3 162 128
AK163269.1 - 2 - 3 339 128
BC032271.1 - 2 - 3 213 4954726 128
EU069461.1 - 2 - 3 146 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0X00002266.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mmgt1

Gene Ontology Categories:

Function(s) Process(es)

transport
cation transport
cobalt ion transport
copper ion transport
ferrous iron transport
magnesium ion transport
ferrous iron transmembrane transport
cobalt ion transmembrane transporter activity
ferrous iron transmembrane transporter activity
magnesium ion transmembrane transporter activity
inorganic cation transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0X00014348.mm.1 NM_146234.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0X00014348.mm.1 NM_146234.3 Refseq
Affymetrix Mouse Clariom S Array PSR0X00014348.mm.1 TC0X00002266.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0