This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00462228_m1
See other CADM4 GE Assays ›
Gene Symbol:
Cadm4
Gene Name:
cell adhesion molecule 4
Gene Aliases:
Igdf4c, Igsf4c, Tsll2
Chromosome Location:
Chr.7: 24482023 - 24504533 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00462228_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cadm4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153112.3 NP_694752.1 8 - 9 1131 68
GenBank mRNA AK143204.1 - 8 - 9 1132 68
AY059394.1 - 8 - 9 1055 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700000384.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mus musculus 1700 Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cadm4

Gene Ontology Categories:

Function(s)

cell adhesion

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005093 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005091 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005092 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005095 - -
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114110 AK143204.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2114110 NM_153112.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 767112 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 265542 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 270935 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 500139 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 603845 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 112241 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 885477 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 187791 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 697438 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 749234 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 227699 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 252519 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 670598 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 111362 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 650363 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 139937 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 12426 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P112788 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114110 AK143204.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2114110 NM_153112.3 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005152 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005215 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005210 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005186 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005217 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005193 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005173 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005178 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005157 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005168 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005153 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005198 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005177 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005176 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005188 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005208 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005179 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005155 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005195 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005211 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005197 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005161 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005201 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005170 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005184 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005175 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005190 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005163 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005218 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005191 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005165 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005205 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005182 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P25005209 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_67_P06141056 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P112788 - -
Whole Mouse Genome G4122A A_51_P477716 AK143204.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P477716 AY059394.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P477716 NM_153112.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P117157 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119494 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P112788 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06540903 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06094265 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06094259 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06094264 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P06540904 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005093 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005092 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005091 - -
Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K A_68_P25005095 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005144 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005103 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005112 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005111 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005139 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005197 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005272 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005195 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005152 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005107 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005189 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005086 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005119 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005247 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005250 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005069 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005110 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005076 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005260 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005186 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005092 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005091 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005120 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005142 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005114 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005074 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005141 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005193 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005262 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005155 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005223 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005182 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005201 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005170 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005090 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005134 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005146 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005126 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005220 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005072 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005251 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005179 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005283 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005211 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005071 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005168 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005123 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005233 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005267 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005088 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005100 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005205 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005063 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005215 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005131 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005278 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005161 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005286 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005237 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005231 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005175 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005184 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005078 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005087 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005117 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005098 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005238 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005243 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005094 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005280 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005274 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005246 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005085 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005258 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005083 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005113 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005265 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005218 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005128 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005097 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005198 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005271 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005109 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005136 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005166 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005208 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005067 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005236 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005217 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005240 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005065 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005254 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005150 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005249 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005191 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P25005226 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005113 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005122 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06094265 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005184 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005093 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06501576 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005208 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005105 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06071329 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06407641 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06629975 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005107 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005084 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06430633 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005076 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005128 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005141 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005215 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005116 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005085 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06660232 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06407640 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005197 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06502869 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06510042 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06128281 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06629971 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06094264 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005086 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06502867 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005126 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005066 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06573207 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005111 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06261522 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06071330 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06343926 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06071322 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06261521 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005110 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06502868 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05890543 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06094268 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005139 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005123 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005182 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005218 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005091 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005131 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005205 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005118 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005202 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005112 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06629964 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005136 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06510043 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P05890544 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06047299 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06161756 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06071324 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005098 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005198 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06094261 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005100 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005134 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005104 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005083 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06343920 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005120 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005090 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06343936 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P25005201 - -
Agilent Six_ChIP_02_of_03_v2 A_68_P06407634 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10550917 - -
Affymetrix Mouse Gene 1.0 ST Array 10550918 - -
Affymetrix Mouse Gene 1.0 ST Array 10550923 - -
Affymetrix Mouse Gene 1.0 ST Array 10550921 - -
Affymetrix Mouse Gene 1.0 ST Array 10550919 - -
Affymetrix Mouse Gene 1.0 ST Array 10550916 - -
Affymetrix Mouse Gene 1.0 ST Array 10550920 - -
Affymetrix Mouse Gene 1.0 ST Array 10550922 - -
Affymetrix Mouse Gene 1.0 ST Array 10550924 - -
Affymetrix HT MG-430 PM Array Plate 1426263_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10550915 - -
Mouse Genome 430 2.0 Array 1426263_at - -
Mouse Genome 430 2.0 Array 1426263_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 260299_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 260299_PM_at - -
Affymetrix Mouse Clariom S Array PSR0700002807.mm.1 TC0700000384.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700002807.mm.1 NM_153112.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700002807.mm.1 NM_153112.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700002808.mm.1 TC0700000384.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700002808.mm.1 NM_153112.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700002808.mm.1 NM_153112.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700002806.mm.1 TC0700000384.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0700002806.mm.1 NM_153112.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700002806.mm.1 NM_153112.3 Refseq
Mouse Expression Set 430 1426263_at - -
Mouse Expression Set 430 1426263_PM_at - -
Mouse Genome 430A 2.0 Array 1426263_PM_at - -
Mouse Genome 430A 2.0 Array 1426263_at - -
Affymetrix HT Mouse Genome 430A Array 1426263_at - -
Affymetrix HT Mouse Genome 430A Array 1426263_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 878864 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 1240377 - -
Illumina mouseRef-8 v1.0 expression beadchip 0001240377 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1223665 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1223665 - -
mouse-6 v1.1 (Illumina) scl32961.9.1_35-S - -
Illumina Sentrix mouse-6.v1 bead array 1240377 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl32961.9.1_35-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 1240377 - -
Illumina Sentrix Mouse-6 Expression BeadChip (3) scl32961.9.1_35-S - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0