This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00462685_m1
See other HSD3B2 GE Assays ›
Gene Symbol:
Hsd3b2
Gene Name:
hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2
Gene Aliases:
-
Chromosome Location:
Chr.3: 98711107 - 98724594 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00462685_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hsd3b2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153193.3 NP_694873.2 3 - 4 209 83
GenBank mRNA BC026757.1 - 3 - 4 209 5136428 83
BC040397.1 - 252 4222081 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0300002558.mm.1

Additional Information:

This assay is in the 5'UTR region of the BC026757.1,BC040397.1,NM_153193.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Mus musculus Mouse Androgen Panel Mouse Estrogen Panel 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hsd3b2

Panther Classification:

Molecular Function -

dehydratase

Gene Ontology Categories:

Function(s) Process(es)

steroid biosynthetic process
metabolic process
oxidation-reduction process
catalytic activity
3-beta-hydroxy-delta5-steroid dehydrogenase activity
steroid delta-isomerase activity
oxidoreductase activity
oxidoreductase activity, acting on the CH-OH group of donors, NAD or NADP as acceptor
isomerase activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023114 BC040397.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023114 M75886.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023114 BC026757.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2023114 NM_153193.3 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036547 BC040397.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036547 BX639392.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036547 M75886.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036547 BC026757.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_55_P2036547 NM_153193.3 mouse_transcriptome
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 107716 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 756606 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 952747 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 407374 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 902539 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 958420 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 403716 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 404844 - -
Agilent SurePrint G3 Mouse CGH Microarray 1x1M 41469 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 52385 - -
Agilent SurePrint G3 Mouse CGH Microarray 4x180K 168382 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P102127 - -
Agilent Mouse Genome CGH Microarray 44K G4426B A_53_P141268 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2132812 - -
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023114 BC040397.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023114 M75886.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023114 BC026757.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2023114 NM_153193.3 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036547 BC040397.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036547 BX639392.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036547 M75886.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036547 BC026757.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_55_P2036547 NM_153193.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226567 BC040397.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226567 BX639392.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226567 M75886.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226567 BC026757.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_51_P226567 NM_153193.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339759 BC040397.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339759 M75886.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339759 BC026757.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P339759 NM_153193.3 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P318447 BC040397.1 mouse_transcriptome
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22360699 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22360701 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22360707 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22360712 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22360706 - -
Agilent Mouse Promoter ChIP-on-Chip A) A_68_P22360693 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P102127 - -
Agilent Mouse Genome CGH Microarray 244A (G4415A) A_53_P141268 - -
Whole Mouse Genome G4122A A_51_P226567 BC040397.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226567 BX639392.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226567 M75886.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226567 BC026757.1 mouse_transcriptome
Whole Mouse Genome G4122A A_51_P226567 NM_153193.3 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P318447 BC040397.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339759 BC040397.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339759 M75886.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339759 BC026757.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P339759 NM_153193.3 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P119411 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P106542 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P141268 - -
Agilent Mouse Genome CGH Microarray 105A (G4416A) A_53_P102127 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02749938 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02566070 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P02865032 - -
Agilent Mouse Proximal Promoter ChIP-on-Chip A) A_68_P03085527 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360601 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360633 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360662 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360664 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360683 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360707 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360656 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360701 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360669 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360612 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360642 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360624 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360693 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360671 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360688 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360634 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360606 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360621 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360636 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360681 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360607 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360603 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360667 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360610 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360654 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360685 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360649 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360617 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360644 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360631 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360706 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360639 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360604 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360665 - -
Agilent Mouse Whole Genome ChIP-on-Chip A_68_P22360599 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360699 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03085531 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360683 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360681 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360686 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02566070 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360690 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P03172862 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360693 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360674 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360706 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02749938 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02476524 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P02556541 - -
Agilent Six_ChIP_01_of_03_v2 A_68_P22360692 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10500553 - -
Affymetrix Mouse Gene 1.0 ST Array 10500552 - -
Affymetrix Mouse Gene 1.0 ST Array 10500549 - -
Affymetrix Mouse Gene 1.0 ST Array 10500550 - -
Affymetrix Mouse Gene 1.0 ST Array 10500548 - -
Affymetrix Mouse Gene 1.0 ST Array 10500551 - -
Affymetrix Mouse Gene 1.0 ST Array 10500554 - -
Affymetrix HT MG-430 PM Array Plate 1425127_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10500547 - -
Mouse Genome 430 2.0 Array 1425127_at - -
Mouse Genome 430 2.0 Array 1425127_PM_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 15493_at - -
Affymetrix GeneChip Mouse Genome 430 2.0 Array 15493_PM_at - -
Affymetrix Mouse Clariom S Array PSR0300019997.mm.1 NM_153193.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019997.mm.1 NM_153193.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300019997.mm.1 TC0300002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300019999.mm.1 NM_153193.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019999.mm.1 NM_153193.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300019999.mm.1 TC0300002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300019996.mm.1 TC0300002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300019998.mm.1 NM_153193.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300019998.mm.1 NM_153193.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300019998.mm.1 TC0300002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300020000.mm.1 NM_153193.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300020000.mm.1 NM_153193.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300020000.mm.1 TC0300002558.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0300020001.mm.1 NM_153193.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0300020001.mm.1 NM_153193.3 Refseq
Affymetrix Mouse Clariom S Array PSR0300020001.mm.1 TC0300002558.mm.1 MTA TC
Mouse Expression Set 430 1460232_s_at - -
Mouse Expression Set 430 1425127_at - -
Mouse Expression Set 430 1425127_PM_at - -
Mouse Expression Set 430 1460232_PM_s_at - -
Murine Genome U74v2 Set 101659_PM_at - -
Murine Genome U74v2 Set 101659_at - -
Mouse Genome 430A 2.0 Array 1460232_s_at - -
Mouse Genome 430A 2.0 Array 1425127_at - -
Mouse Genome 430A 2.0 Array 1425127_PM_at - -
Mouse Genome 430A 2.0 Array 1460232_PM_s_at - -
Affymetrix HT Mouse Genome 430A Array 1425127_PM_at - -
Affymetrix HT Mouse Genome 430A Array 1425127_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 629739 - -
Array Name Probe IDs Targets Target Type
Illumina mouse-6 v1.0 expression beadchip 5690333 - -
Illumina mouse-6 v1.0 expression beadchip 70068 - -
Illumina mouse-6 v1.0 expression beadchip 4230170 - -
Illumina mouse-6 v1.0 expression beadchip 4120114 - -
Illumina mouse-6 v1.0 expression beadchip 770037 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004230170 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000770037 - -
Illumina mouseRef-8 v1.0 expression beadchip 0004120114 - -
Illumina mouseRef-8 v1.0 expression beadchip 0005690333 - -
Illumina mouseRef-8 v1.0 expression beadchip 0000070068 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1214838 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1241116 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1233611 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1259238 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_1234480 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1234480 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1233611 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1241116 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1259238 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_1214838 - -
mouse-6 v1.1 (Illumina) scl0015493.2_83-S - -
mouse-6 v1.1 (Illumina) scl0015493.2_185-S - -
mouse-6 v1.1 (Illumina) scl015493.1_155-S - -
mouse-6 v1.1 (Illumina) scl21777.6_80-S - -
mouse-6 v1.1 (Illumina) scl0015493.1_84-S - -
Illumina Sentrix mouse-6.v1 bead array 70068 - -
Illumina Sentrix mouse-6.v1 bead array 770037 - -
Illumina Sentrix mouse-6.v1 bead array 4230170 - -
Illumina Sentrix mouse-6.v1 bead array 5690333 - -
Illumina Sentrix mouse-6.v1 bead array 4120114 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl015493.1_155-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl21777.6_80-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0015493.2_83-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0015493.2_185-S - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl0015493.1_84-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 770037 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4120114 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 5690333 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 70068 - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 4230170 - -
MouseRef-8 v2 Expression BeadChip ILMN_1234480 - -
MouseRef-8 v2 Expression BeadChip ILMN_1241116 - -
MouseRef-8 v2 Expression BeadChip ILMN_1214838 - -
MouseRef-8 v2 Expression BeadChip ILMN_1259238 - -
MouseRef-8 v2 Expression BeadChip ILMN_1233611 - -
MouseRef-8 v2 Expression BeadChip ILMN_2898958 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1214838 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1233611 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1259238 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2898958 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1241116 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_1234480 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0