This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00462850_m1
See other MON2 GE Assays ›
Gene Symbol:
Mon2
Gene Name:
MON2 homolog (yeast)
Gene Aliases:
2610528O22Rik, AW495628, Sf21, mKIAA1040
Chromosome Location:
Chr.10: 122992060 - 123076505 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00462850_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mon2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001163024.1 NP_001156496.1 4 - 5 982 74
NM_001163025.1 NP_001156497.1 4 - 5 982 74
NM_153395.2 NP_700444.2 4 - 5 982 74
GenBank mRNA AB014059.1 - 4 - 5 504 74
AK122425.1 - 4 - 5 982 74
AK161217.1 - 4 - 5 861 74
BC064131.1 - 4 - 5 472 74
BC150943.1 - 4 - 5 503 9087854 74
BE955337.1 - 2 - 3 180 74

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mon2

Panther Classification:

Molecular Function -

guanyl-nucleotide exchange factor

Gene Ontology Categories:

Function(s) Process(es)

transport
protein transport
vesicle-mediated transport
positive regulation of GTPase activity
ARF guanyl-nucleotide exchange factor activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0