This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00463554_m1
See other MBOAT1 GE Assays ›
Gene Symbol:
Mboat1
Gene Name:
membrane bound O-acyltransferase domain containing 1
Gene Aliases:
9130215M02Rik, BC023845, LPEAT1, Moact1, Oact1
Chromosome Location:
Chr.13: 30136449 - 30246694 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00463554_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mboat1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153546.4 NP_705774.1 8 - 9 1084 68
GenBank mRNA AB297382.1 - 8 - 9 907 68
AB305045.1 - 8 - 9 907 68
AK033388.1 - 8 - 9 1026 68
AK137755.1 - 8 - 9 1017 68
AK146984.1 - 8 - 9 991 68
BC023845.1 - 8 - 9 1088 5346000 68
BC024653.1 - 8 - 9 1036 3660479 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1300000330.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mboat1

Panther Classification:

Molecular Function -

acetyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
biological_process
phospholipid biosynthetic process
molecular_function
dihydrolipoamide branched chain acyltransferase activity
sterol O-acyltransferase activity
N-acetyltransferase activity
O-acyltransferase activity
palmitoleoyl [acyl-carrier-protein]-dependent acyltransferase activity
carnitine O-acyltransferase activity
acetyltransferase activity
C-acyltransferase activity
palmitoyltransferase activity
N-acyltransferase activity
acylglycerol O-acyltransferase activity
serine O-acyltransferase activity
O-acetyltransferase activity
O-octanoyltransferase activity
octanoyltransferase activity
O-palmitoyltransferase activity
S-acyltransferase activity
S-acetyltransferase activity
S-malonyltransferase activity
malonyltransferase activity
C-palmitoyltransferase activity
transferase activity
transferase activity, transferring acyl groups
succinyltransferase activity
N-succinyltransferase activity
O-succinyltransferase activity
S-succinyltransferase activity
sinapoyltransferase activity
O-sinapoyltransferase activity
peptidyl-lysine N6-myristoyltransferase activity
peptidyl-lysine N6-palmitoyltransferase activity
benzoyl acetate-CoA thiolase activity
3-hydroxybutyryl-CoA thiolase activity
3-ketopimelyl-CoA thiolase activity
N-palmitoyltransferase activity
myristoyltransferase activity
acyl-CoA N-acyltransferase activity
protein-cysteine S-myristoyltransferase activity
protein-cysteine S-acyltransferase activity
dihydrolipoamide S-acyltransferase activity
glucosaminyl-phosphotidylinositol O-acyltransferase activity
ergosterol O-acyltransferase activity
lanosterol O-acyltransferase activity
naphthyl-2-oxomethyl-succinyl-CoA succinyl transferase activity
2,4,4-trimethyl-3-oxopentanoyl-CoA 2-C-propanoyl transferase activity
2-methylhexanoyl-CoA C-acetyltransferase activity
butyryl-CoA 2-C-propionyltransferase activity
2,6-dimethyl-5-methylene-3-oxo-heptanoyl-CoA C-acetyltransferase activity
L-2-aminoadipate N-acetyltransferase activity
UDP-3-O-[3-hydroxymyristoyl] glucosamine N-acyltransferase activity
keto acid formate lyase activity
Ras palmitoyltransferase activity
azetidine-2-carboxylic acid acetyltransferase activity
peptidyl-lysine N-acetyltransferase activity, acting on acetyl phosphate as donor
acetyl-CoA:L-lysine N6-acetyltransferase

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1300002004.mm.1 NM_153546.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1300002004.mm.1 NM_153546.4 Refseq
Affymetrix Mouse Clariom S Array PSR1300002004.mm.1 TC1300000330.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0