This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00464217_g1
See other POLR2B GE Assays ›
Gene Symbol:
Polr2b
Gene Name:
polymerase (RNA) II (DNA directed) polypeptide B
Gene Aliases:
Pol2rb, Rpb140, Rpb2
Chromosome Location:
Chr.5: 77310484 - 77349328 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00464217_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Polr2b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153798.2 NP_722493.2 19 - 20 2896 79
GenBank mRNA AK039766.1 - 19 - 20 2877 79
BC016415.1 - 19 - 20 2882 79
BC038472.1 - 19 - 20 2889 5026936 79
CF553167.1 - 2 - 3 335 79

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Polr2b

Panther Classification:

Molecular Function -

DNA-directed RNA polymerase

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
transcription from RNA polymerase II promoter
RNA polymerase II activity
DNA binding
chromatin binding
DNA-directed RNA polymerase activity
transferase activity
nucleotidyltransferase activity
ribonucleoside binding
poly(A) RNA binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0