This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00464359_m1
See other IGSF11 GE Assays ›
Gene Symbol:
Igsf11
Gene Name:
immunoglobulin superfamily, member 11
Gene Aliases:
1700025L02Rik, BT-IgSF, Igsf13
Chromosome Location:
Chr.16: 38892476 - 39027159 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00464359_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Igsf11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_170599.2 NP_733548.2 3 - 4 585 94
GenBank mRNA AB079880.1 - 3 - 4 422 94
AK160146.1 - 3 - 4 548 94
BC057555.1 - 3 - 4 585 6416190 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000564.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Igsf11

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin

Gene Ontology Categories:

Function(s)

cell adhesion
regulation of growth

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600005350.mm.1 NM_170599.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600005350.mm.1 NM_170599.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600005350.mm.1 TC1600000564.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0