This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00465075_m1
See other PRKDC GE Assays ›
Gene Symbol:
Prkdc
Gene Name:
protein kinase, DNA activated, catalytic polypeptide
Gene Aliases:
AI326420, AU019811, DNA-PKcs, DNAPDcs, DNAPK, DNPK1, DOXNPH, HYRC1, XRCC7, dxnph, p460, scid, slip
Chromosome Location:
Chr.16: 15637444 - 15842252 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00465075_m1
Size
Availability Made To Order
Catalog # 4351372
Price 417.00
Your Price
Online offer: 387.65
387.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Prkdc
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_011159.2 NP_035289.2 11 - 12 1131 84
GenBank mRNA AB007544.1 - 11 - 12 1108 84
AB011543.1 - 11 - 12 1120 84
AK084827.1 - 11 - 12 1132 84
D87521.1 - 11 - 12 1108 84

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Prkdc

Panther Classification:

Molecular Function -

non-receptor serine/threonine protein kinase protein modifying enzyme

Gene Ontology Categories:

Function(s) Process(es)

telomere maintenance
double-strand break repair via homologous recombination
somitogenesis
negative regulation of protein phosphorylation
B cell lineage commitment
pro-B cell differentiation
T cell lineage commitment
immunoglobulin production
negative regulation of immunoglobulin production
positive regulation of immune system process
DNA repair
double-strand break repair
double-strand break repair via nonhomologous end joining
protein phosphorylation
cellular response to DNA damage stimulus
brain development
heart development
intrinsic apoptotic signaling pathway in response to DNA damage
response to ionizing radiation
response to gamma radiation
phosphorylation
peptidyl-serine phosphorylation
lymphocyte differentiation
protein destabilization
cellular response to insulin stimulus
T cell differentiation in thymus
V(D)J recombination
immunoglobulin V(D)J recombination
T cell receptor V(D)J recombination
ectopic germ cell programmed cell death
regulation of circadian rhythm
positive regulation of apoptotic process
negative regulation of apoptotic process
positive regulation of transcription from RNA polymerase II promoter
positive regulation of fibroblast proliferation
rhythmic process
spleen development
thymus development
positive regulation of developmental growth
signal transduction involved in mitotic G1 DNA damage checkpoint
negative regulation of cellular senescence
negative regulation of response to gamma radiation
nucleotide binding
DNA binding
double-stranded DNA binding
protein kinase activity
protein serine/threonine kinase activity
DNA-dependent protein kinase activity
protein binding
ATP binding
transcription factor binding
kinase activity
transferase activity
transferase activity, transferring phosphorus-containing groups
phosphotransferase activity, alcohol group as acceptor
enzyme binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-blq6p:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline