This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00466106_m1
See other DISP1 GE Assays ›
Gene Symbol:
Disp1
Gene Name:
dispatched homolog 1 (Drosophila)
Gene Aliases:
1190008H24Rik, DispA
Chromosome Location:
Chr.1: 183086264 - 183221536 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00466106_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Disp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278218.1 NP_001265147.1 2 - 3 336 53
NM_026866.3 NP_081142.3 3 - 4 424 53
GenBank mRNA AK138276.1 - 3 - 4 381 53
AY150698.1 - 3 - 4 316 53
BC043102.1 - 2 - 3 297 6406376 53
BC059225.1 - 2 - 3 297 6406376 53

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK138276.1, AY150698.1, BC043102.1, BC059225.1, NM_001278218.1, NM_026866.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Disp1

Gene Ontology Categories:

Function(s) Process(es)

smoothened signaling pathway
patched ligand maturation
multicellular organism development
determination of left/right symmetry
pattern specification process
embryonic pattern specification
dorsal/ventral pattern formation
peptide transport
diaphragm development
hedgehog receptor activity
peptide transporter activity

MicroArray Probes:

Agilent Affymetrix ABI Illumina

Array Name Probe IDs Targets Target Type
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AK138276.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AK170113.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AY144589.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AY150577.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AY150698.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AK138276.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AK170113.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AY144589.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AY150577.1 mouse_transcriptome
Agilent Whole Mouse Genome Microarray 4x44K v2 A_52_P377416 AY150698.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AK138276.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AK170113.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AY144589.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AY150577.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AY150698.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AK138276.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AK170113.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AY144589.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AY150577.1 mouse_transcriptome
SurePrint G3 Mouse Gene Expression Microarray G4852A A_52_P377416 AY150698.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P243152 AK138276.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P243152 AK004521.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P243152 AK170113.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P243152 AY144589.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P243152 AY150577.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AK138276.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AK170113.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AY144589.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AY150577.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AY150698.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AK138276.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AK170113.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AY144589.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AY150577.1 mouse_transcriptome
Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " A_52_P377416 AY150698.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AK138276.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AK170113.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AY144589.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AY150577.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AY150698.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AK138276.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AK170113.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AY144589.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AY150577.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P377416 AY150698.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243152 AK138276.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243152 AK004521.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243152 AK170113.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243152 AY144589.1 mouse_transcriptome
Whole Mouse Genome G4122A A_52_P243152 AY150577.1 mouse_transcriptome
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P108667 - -
Agilent Mouse Developmental Microarray 44K G2519A Option 002 A_66_P100639 - -
Array Name Probe IDs Targets Target Type
Affymetrix Mouse Gene 1.0 ST Array 10468912 - -
Affymetrix Mouse Gene 1.0 ST Array 10468915 - -
Affymetrix Mouse Gene 1.0 ST Array 10598179 - -
Affymetrix Mouse Gene 1.0 ST Array 10468913 - -
Affymetrix Mouse Gene 1.0 ST Array 10468911 - -
Affymetrix Mouse Gene 1.0 ST Array 10468910 - -
Affymetrix Mouse Gene 1.0 ST Array 10468914 - -
Affymetrix HT MG-430 PM Array Plate 1434795_PM_at - -
Affymetrix HT MG-430 PM Array Plate 1435132_PM_at - -
Affymetrix Mouse Gene 1.1 ST Array 10598178 - -
Affymetrix Mouse Gene 1.1 ST Array 10468909 - -
Mouse Genome 430 2.0 Array 1435132_PM_at - -
Mouse Genome 430 2.0 Array 1434795_PM_at - -
Mouse Genome 430 2.0 Array 1435132_at - -
Mouse Genome 430 2.0 Array 1434795_at - -
Mouse Expression Set 430 1435132_at - -
Mouse Expression Set 430 1434795_at - -
Mouse Expression Set 430 1434795_PM_at - -
Mouse Expression Set 430 1435132_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1434795_at - -
Affymetrix HT Mouse Genome 430B Array 1435132_at - -
Affymetrix HT Mouse Genome 430B Array 1435132_PM_at - -
Affymetrix HT Mouse Genome 430B Array 1434795_PM_at - -
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 369182 - -
Array Name Probe IDs Targets Target Type
MouseWG-6 v2 Expression BeadChip 3780195 - -
Illumina mouse-6 v1.0 expression beadchip 3780195 - -
Illumina mouseRef-8 v1.0 expression beadchip 0003780195 - -
Illumina mouseRef-8 v1.1 expression beadchip ILMN_2772288 - -
Illumina mouse-6 v1.1 expression beadchip ILMN_2772288 - -
mouse-6 v1.1 (Illumina) scl068897.1_240-S - -
Illumina Sentrix mouse-6.v1 bead array 3780195 - -
Illumina Sentrix MouseRef-8 Expression BeadChip scl068897.1_240-S - -
Illumina Sentrix Mouse-6 Expression BeadChip (2) 3780195 - -
MouseRef-8 v2 Expression BeadChip ILMN_2772288 - -
Illumina MouseWG-6 v2.0 expression beadchip ILMN_2772288 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0