This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00466110_m1
See other DISP1 GE Assays ›
Gene Symbol:
Disp1
Gene Name:
dispatched homolog 1 (Drosophila)
Gene Aliases:
1190008H24Rik, DispA
Chromosome Location:
Chr.1: 183086264 - 183221536 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00466110_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Disp1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278218.1 NP_001265147.1 7 - 8 1238 94
NM_001278219.1 NP_001265148.1 7 - 8 1198 94
NM_001278220.1 NP_001265149.1 6 - 7 1081 94
NM_026866.3 NP_081142.3 8 - 9 1326 94
GenBank mRNA AK004521.1 - 6 - 7 1056 94
AK138276.1 - 8 - 9 94
AK170113.1 - 7 - 8 1193 94
AY144589.1 - 6 - 7 1036 94
AY150577.1 - 6 - 7 985 94
AY150698.1 - 8 - 9 1218 94
BC043102.1 - 7 - 8 1199 6406376 94
BC059225.1 - 7 - 8 1199 6406376 94

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003721.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Disp1

Gene Ontology Categories:

Function(s) Process(es)

smoothened signaling pathway
patched ligand maturation
multicellular organism development
determination of left/right symmetry
pattern specification process
embryonic pattern specification
dorsal/ventral pattern formation
peptide transport
diaphragm development
hedgehog receptor activity
peptide transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100030479.mm.1 NM_001278218.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100030479.mm.1 NM_001278219.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100030479.mm.1 NM_001278220.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100030479.mm.1 NM_026866.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100030479.mm.1 TC0100003721.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0