This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00466469_m1
See other RBM12 GE Assays ›
Gene Symbol:
Rbm12
Gene Name:
RNA binding motif protein 12
Gene Aliases:
5730420G12Rik, 9430070C08Rik, AI852903, SWAN, mKIAA0765
Chromosome Location:
Chr.2: 156094882 - 156111965 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00466469_m1
Size
Availability Inventoried
Catalog # 4331182
Price 284.00
Your Price
Online offer: 266.65
266.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Rbm12
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_029397.3 NP_083673.3 1 - 2 181 57
GenBank mRNA AF393215.1 - 1 - 2 144 57
AK035014.1 - 1 - 2 139 57
AK080772.1 - 1 - 2 139 57
BC052473.1 - 1 - 2 124 6833669 57

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0200005511.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF393215.1,AK035014.1,AK080772.1,BC052473.1,NM_029397.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Rbm12

Panther Classification:

Molecular Function -

RNA splicing factor

Gene Ontology Categories:

Function(s) Process(es)

biological_process
nucleotide binding
nucleic acid binding
RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 NM_029397.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 NM_029397.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 NM_170588.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 NM_170588.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 NM_170598.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 NM_170598.2 Refseq
Affymetrix Mouse Clariom S Array PSR0200042493.mm.1 TC0200005511.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0200042494.mm.1 NM_029397.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0200042494.mm.1 NM_029397.3 Refseq
Affymetrix Mouse Clariom S Array PSR0200042494.mm.1 TC0200005511.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 921189 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0