This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00467609_g1
See other NDUFS2 GE Assays ›
Gene Symbol:
Ndufs2
Gene Name:
NADH dehydrogenase (ubiquinone) Fe-S protein 2
Gene Aliases:
AL033311
Chromosome Location:
Chr.1: 171234860 - 171247112 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00467609_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufs2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_153064.4 NP_694704.1 10 - 11 1234 71
GenBank mRNA AK078474.1 - 10 - 11 1235 71
AK146269.1 - 10 - 11 1239 71
AK150431.1 - 10 - 11 1230 71
AK151746.1 - 10 - 11 1207 71
AK153364.1 - 10 - 11 1207 71
AK165426.1 - 10 - 11 1230 71
AK167016.1 - 10 - 11 1411 71
AK168971.1 - 11 - 12 1205 71
AK209966.1 - 1 - 2 93 71
BC003898.1 - 10 - 11 1233 71
BC016097.1 - 10 - 11 1221 4910260 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003534.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufs2

Panther Classification:

Molecular Function -

oxidoreductase

Gene Ontology Categories:

Function(s) Process(es)

transport
response to oxidative stress
oxidation-reduction process
NADH dehydrogenase activity
NADH dehydrogenase (ubiquinone) activity
oxidoreductase activity
oxidoreductase activity, acting on NAD(P)H
ubiquitin protein ligase binding
metal ion binding
quinone binding
NAD binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100028918.mm.1 NM_153064.5 Refseq
Affymetrix Mouse Clariom S Array PSR0100028918.mm.1 TC0100003534.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-z4mpm:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0