This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00468417_g1
See other EWSR1 GE Assays ›
Gene Symbol:
Ewsr1
Gene Name:
Ewing sarcoma breakpoint region 1
Gene Aliases:
Ews, Ewsh
Chromosome Location:
Chr.11: 5069687 - 5099099 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00468417_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ewsr1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001283061.1 NP_001269990.1 7 - 8 865 75
NM_001283062.1 NP_001269991.1 7 - 8 865 75
NM_001283063.1 NP_001269992.1 7 - 8 865 75
NM_007968.3 NP_031994.2 7 - 8 865 75
GenBank mRNA AK019460.1 - 7 - 8 814 75
AK034755.1 - 7 - 8 841 75
AK049743.1 - 7 - 8 845 75
AK146791.1 - 7 - 8 841 75
AK147103.1 - 7 - 8 832 75
AK147909.1 - 7 - 8 822 75
AK150345.1 - 7 - 8 823 75
AK151625.1 - 7 - 8 836 75
AK153276.1 - 7 - 8 823 75
BC068226.1 - 7 - 8 832 5707017 75
X79233.1 - 7 - 8 854 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ewsr1

Panther Classification:

Molecular Function -

RNA metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
nucleotide binding
nucleic acid binding
RNA binding
protein binding
calmodulin binding
zinc ion binding
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0