This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00468777_m1
See other EXT2 GE Assays ›
Gene Symbol:
Ext2
Gene Name:
exostoses (multiple) 2
Gene Aliases:
AI893565
Chromosome Location:
Chr.2: 93695631 - 93822588 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00468777_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ext2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010163.3 NP_034293.2 5 - 6 1155 112
GenBank mRNA AK082572.1 - 5 - 6 1137 112
AK134438.1 - 1 - 2 96 112
AK148090.1 - 5 - 6 1144 113
AK148369.1 - 4 - 5 916 112
AK150155.1 - 5 - 6 1147 112
AK152265.1 - 1255 112
AK152840.1 - 1257 112
AK164342.1 - 5 - 6 1141 112
BC006597.1 - 5 - 6 1128 3965079 112
U67837.1 - 5 - 6 1130 112
U72141.1 - 5 - 6 1134 112

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ext2

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

ossification
mesoderm formation
glycosaminoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process
heparan sulfate proteoglycan biosynthetic process, polysaccharide chain biosynthetic process
cell differentiation
cellular polysaccharide biosynthetic process
acetylglucosaminyltransferase activity
glucuronosyltransferase activity
transferase activity
transferase activity, transferring glycosyl groups
heparan sulfate N-acetylglucosaminyltransferase activity
protein homodimerization activity
metal ion binding
protein heterodimerization activity
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity
N-acetylglucosaminyl-proteoglycan 4-beta-glucuronosyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0