This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00468921_m1
See other HMOX2 GE Assays ›
Gene Symbol:
Hmox2
Gene Name:
heme oxygenase 2
Gene Aliases:
HO-2, HO2
Chromosome Location:
Chr.16: 4726361 - 4766249 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00468921_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hmox2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_010443.2 NP_034573.2 1 - 2 124 107
GenBank mRNA AF029874.1 - 1 - 2 121 107
AF054669.1 - 1 - 2 125 107
CA465845.1 - 1 - 2 139 107

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1600000033.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF029874.1,AF054669.1,NM_010443.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hmox2

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

response to hypoxia
heme oxidation
response to oxidative stress
oxidation-reduction process
heme oxygenase (decyclizing) activity
oxidoreductase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1600000267.mm.1 NM_010443.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000267.mm.1 NM_010443.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000267.mm.1 TC1600000033.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1600000274.mm.1 NM_001136066.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000274.mm.1 NM_001136066.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000274.mm.1 NM_010443.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1600000274.mm.1 NM_010443.2 Refseq
Affymetrix Mouse Clariom S Array PSR1600000274.mm.1 TC1600000033.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0