This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469043_g1
See other EIF4G2 GE Assays ›
Gene Symbol:
Eif4g2
Gene Name:
eukaryotic translation initiation factor 4, gamma 2
Gene Aliases:
AA589388, DAP-5, E130105L11Rik, Nat1, Natm1, p97
Chromosome Location:
Chr.7: 111067985 - 111083030 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469043_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Eif4g2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001040131.2 NP_001035221.1 11 - 12 1319 74
NM_013507.3 NP_038535.2 11 - 12 1319 74
GenBank mRNA AK141512.1 - 4 - 5 389 74
AK144309.1 - 11 - 12 1293 74
AK150745.1 - 11 - 12 1296 74
AK151637.1 - 11 - 12 1296 74
BC039274.1 - 10 - 11 997 74
BC040391.1 - 10 - 11 1008 4018962 74
BC043034.1 - 10 - 11 999 5696921 74
BC056387.1 - 11 - 12 1283 6854755 74
BC057673.1 - 12 - 13 1171 4020759 74
BC064810.1 - 10 - 11 1052 6825556 74
BC092521.1 - 11 - 12 1274 30649919 74
BC127064.1 - 11 - 12 1319 40109978 74
BU610672.1 - 3 - 4 301 74
U63323.1 - 11 - 12 1271 74
U76112.1 - 11 - 12 1293 74

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0700004108.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Eif4g2

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

translation
translational initiation
regulation of translation
regulation of translational initiation
RNA binding
translation initiation factor activity
protein binding
translation factor activity, RNA binding
poly(A) RNA binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0700034067.mm.1 NM_001040131.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034067.mm.1 NM_001040131.2 Refseq
Affymetrix Mouse Clariom S Array PSR0700034067.mm.1 NM_013507.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0700034067.mm.1 NM_013507.3 Refseq
Affymetrix Mouse Clariom S Array PSR0700034067.mm.1 TC0700004108.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4krp5:80/100.66.77.213:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0