This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469124_g1
See other DMTN GE Assays ›
Gene Symbol:
Dmtn
Gene Name:
dematin actin binding protein
Gene Aliases:
AI325486, Epb4.9, Epb49, dematin
Chromosome Location:
Chr.14: 70602184 - 70636048 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469124_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Dmtn
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001252662.1 NP_001239591.1 7 - 8 935 67
NM_001252663.1 NP_001239592.1 7 - 8 935 67
NM_001252664.1 NP_001239593.1 6 - 7 762 67
NM_001252665.1 NP_001239594.1 6 - 7 661 67
NM_001252666.1 NP_001239595.1 5 - 6 694 67
NM_013514.4 NP_038542.1 8 - 9 876 67
GenBank mRNA AF079846.3 - 6 - 7 605 67
AF155547.1 - 6 - 7 605 67
AK143840.1 - 7 - 8 929 67
AK149301.1 - 6 - 7 572 67
AK155927.1 - 7 - 8 930 67
AK158744.1 - 6 - 7 693 67
AK162866.1 - 6 - 7 613 67
AK165204.1 - 928 67
AK165294.1 - 7 - 8 927 67
BC016897.1 - 8 - 9 876 4240747 67
BC037021.1 - 5 - 6 681 5366040 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400002346.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Dmtn

Panther Classification:

Molecular Function -

actin or actin-binding cytoskeletal protein

Gene Ontology Categories:

Function(s) Process(es)

protein complex assembly
cytoskeleton organization
regulation of cell shape
regulation of lamellipodium assembly
positive regulation of fibroblast migration
negative regulation of peptidyl-threonine phosphorylation
negative regulation of cell-substrate adhesion
actin cytoskeleton organization
positive regulation of blood coagulation
regulation of actin cytoskeleton organization
negative regulation of peptidyl-serine phosphorylation
calcium-mediated signaling using intracellular calcium source
calcium-mediated signaling using extracellular calcium source
erythrocyte development
negative regulation of peptidyl-tyrosine phosphorylation
actin filament bundle assembly
regulation of filopodium assembly
actin filament capping
negative regulation of focal adhesion assembly
protein secretion by platelet
cellular response to calcium ion
cellular response to cAMP
positive regulation of wound healing
negative regulation of protein targeting to membrane
actin filament reorganization
negative regulation of substrate adhesion-dependent cell spreading
positive regulation of substrate adhesion-dependent cell spreading
positive regulation of platelet aggregation
positive regulation of integrin-mediated signaling pathway
actin binding
receptor binding
spectrin binding
protein self-association

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252662.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252662.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252663.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252663.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252664.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252664.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252665.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252665.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252666.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_001252666.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_013514.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 NM_013514.4 Refseq
Affymetrix Mouse Clariom S Array PSR1400018355.mm.1 TC1400002346.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0