This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469149_g1
See other PTPN6 GE Assays ›
Gene Symbol:
Ptpn6
Gene Name:
protein tyrosine phosphatase, non-receptor type 6
Gene Aliases:
70Z-SHP, Hcph, PTPTY-42, Ptp1C, SH-PTP1, SHP-1, hcp, me
Chromosome Location:
Chr.6: 124720707 - 124738709 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469149_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ptpn6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001077705.2 NP_001071173.1 5 - 6 834 59
NM_013545.3 NP_038573.2 5 - 6 832 59
GenBank mRNA AK132509.1 - 5 - 6 751 59
AK150509.1 - 5 - 6 739 59
AK151077.1 - 5 - 6 753 59
AK151623.1 - 5 - 6 756 59
AK159686.1 - 5 - 6 791 59
AK159833.1 - 5 - 6 784 59
AK167923.1 - 5 - 6 786 59
AK169804.1 - 5 - 6 834 59
M68902.1 - 5 - 6 784 59
M90389.1 - 5 - 6 734 59

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0600003114.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Protein Phosphatase Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ptpn6

Panther Classification:

Molecular Function -

protein phosphatase

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
negative regulation of humoral immune response mediated by circulating immunoglobulin
protein dephosphorylation
signal transduction
cell proliferation
positive regulation of cell proliferation
positive regulation of phosphatidylinositol 3-kinase signaling
dephosphorylation
peptidyl-tyrosine phosphorylation
cytokine-mediated signaling pathway
cell differentiation
platelet formation
abortive mitotic cell cycle
positive regulation of cell adhesion mediated by integrin
peptidyl-tyrosine dephosphorylation
intracellular signal transduction
megakaryocyte development
negative regulation of T cell proliferation
natural killer cell mediated cytotoxicity
negative regulation of MAP kinase activity
negative regulation of MAPK cascade
regulation of B cell differentiation
negative regulation of peptidyl-tyrosine phosphorylation
B cell receptor signaling pathway
negative regulation of B cell receptor signaling pathway
negative regulation of T cell receptor signaling pathway
regulation of release of sequestered calcium ion into cytosol
regulation of ERK1 and ERK2 cascade
platelet aggregation
regulation of G1/S transition of mitotic cell cycle
phosphotyrosine binding
phosphoprotein phosphatase activity
protein tyrosine phosphatase activity
transmembrane receptor protein tyrosine phosphatase activity
protein binding
hydrolase activity
phosphatase activity
SH3 domain binding
protein kinase binding
SH2 domain binding
natural killer cell lectin-like receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0600024092.mm.1 TC0600003114.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0600024092.mm.1 NM_001077705.2 Refseq
Affymetrix Mouse Clariom S Array PSR0600024092.mm.1 NM_013545.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hk65g:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0