This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00469326_g1
See other ECH1 GE Assays ›
Gene Symbol:
Ech1
Gene Name:
enoyl coenzyme A hydratase 1, peroxisomal
Gene Aliases:
AA617331
Chromosome Location:
Chr.7: 28824804 - 28832248 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00469326_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ech1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_016772.1 NP_058052.1 7 - 8 680 83
GenBank mRNA AF030343.1 - 7 - 8 680 83
AK088495.1 - 7 - 8 668 83
BC068112.1 - 7 - 8 680 5710971 83
BC087924.1 - 7 - 8 741 6706361 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ech1

Panther Classification:

Molecular Function -

hydratase lyase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
fatty acid metabolic process
metabolic process
C-8 sterol isomerase activity
catalytic activity
receptor binding
delta5-delta2,4-dienoyl-CoA isomerase activity
isomerase activity
3-carboxymuconate cycloisomerase type II activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0