This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00470422_m1
See other MXRA8 GE Assays ›
Gene Symbol:
Mxra8
Gene Name:
matrix-remodelling associated 8
Gene Aliases:
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam
Chromosome Location:
Chr.4: 155839680 - 155844102 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00470422_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mxra8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024263.4 NP_077225.4 1 - 2 210 100
GenBank mRNA AB040488.1 - 1 - 2 130 100
AB052097.1 - 1 - 2 88 100
AK004732.1 - 1 - 2 93 100
BC026438.1 - 1 - 2 132 4239626 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002095.mm.1

Additional Information:

This assay is in the 5'UTR region of the AB040488.1,AB052097.1,AK004732.1,BC026438.1,NM_024263.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mxra8

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017482.mm.1 NM_024263.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017482.mm.1 NM_024263.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400017482.mm.1 TC0400002095.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017480.mm.1 NM_024263.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017480.mm.1 NM_024263.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400017480.mm.1 TC0400002095.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017481.mm.1 NM_024263.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017481.mm.1 NM_024263.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400017481.mm.1 TC0400002095.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0400017483.mm.1 NM_024263.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017483.mm.1 NM_024263.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400017483.mm.1 TC0400002095.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 782274 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-hhcgv:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0