This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00470425_g1
See other MXRA8 GE Assays ›
Gene Symbol:
Mxra8
Gene Name:
matrix-remodelling associated 8
Gene Aliases:
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam
Chromosome Location:
Chr.4: 155839680 - 155844102 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00470425_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mxra8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024263.4 NP_077225.4 5 - 6 1105 114
GenBank mRNA AB040488.1 - 5 - 6 1025 114
AB052097.1 - 5 - 6 983 114
AK004732.1 - 5 - 6 988 114
BC026438.1 - 5 - 6 1027 4239626 114
EU849596.1 - 5 - 6 943 114
EU849597.1 - 5 - 6 934 114

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002095.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mxra8

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017496.mm.1 NM_024263.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017496.mm.1 NM_024263.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400017496.mm.1 TC0400002095.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0