This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00470428_g1
See other MXRA8 GE Assays ›
Gene Symbol:
Mxra8
Gene Name:
matrix-remodelling associated 8
Gene Aliases:
1200013A08Rik, 1700095D18Rik, AI131686, Asp3, Dicam
Chromosome Location:
Chr.4: 155839680 - 155844102 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00470428_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Mxra8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_024263.4 NP_077225.4 8 - 9 1379 72
GenBank mRNA AB040488.1 - 8 - 9 1299 72
AB052097.1 - 8 - 9 1257 72
AK004732.1 - 8 - 9 1262 72
BC002140.1 - 3 - 4 376 72
BC026438.1 - 8 - 9 1301 4239626 72
EU849596.1 - 8 - 9 1217 72
EU849597.1 - 8 - 9 1208 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0400002095.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Mxra8

Gene Ontology Categories:

Function(s) Process(es)

biological_process
molecular_function

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0400017502.mm.1 NM_024263.4.1 Refseq
Affymetrix Mouse Clariom S Array PSR0400017502.mm.1 NM_024263.4 Refseq
Affymetrix Mouse Clariom S Array PSR0400017502.mm.1 TC0400002095.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0