This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00470611_m1
See other HOGA1 GE Assays ›
Gene Symbol:
Hoga1
Gene Name:
4-hydroxy-2-oxoglutarate aldolase 1
Gene Aliases:
0610010D20Rik, Dhdpsl, Npl2
Chromosome Location:
Chr.19: 42045851 - 42070939 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00470611_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Hoga1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026152.1 NP_080428.1 3 - 4 522 64
GenBank mRNA AK002457.1 - 3 - 4 522 64
AK010857.1 - 3 - 4 482 64
BC016430.1 - 3 - 4 491 4235003 64

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000607.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Hoga1

Panther Classification:

Molecular Function -

lyase

Gene Ontology Categories:

Function(s) Process(es)

metabolic process
glyoxylate catabolic process
4-hydroxyproline catabolic process
oxalate metabolic process
pyruvate biosynthetic process
glyoxylate metabolic process
catalytic activity
4-hydroxy-2-oxoglutarate aldolase activity
lyase activity
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900005432.mm.1 NM_026152.1.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005432.mm.1 NM_026152.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900005432.mm.1 TC1900000607.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0