This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00470660_m1
See other PNPLA8 GE Assays ›
Gene Symbol:
Pnpla8
Gene Name:
patatin-like phospholipase domain containing 8
Gene Aliases:
1200006O19Rik, AI467579, Ipla2(gamma)
Chromosome Location:
Chr.12: 44269128 - 44315935 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00470660_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnpla8
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026164.2 NP_080440.2 8 - 9 2100 83
GenBank mRNA AB044139.1 - 8 - 9 1979 83
AK004621.1 - 8 - 9 2067 83
AK017838.1 - 2 - 3 217 83
AK084928.1 - 1767 83
AK145776.1 - 8 - 9 2010 83
AK163211.1 - 8 - 9 2068 83
AK168475.1 - 8 - 9 2063 83
BC019364.1 - 5 - 6 554 83
BC127056.1 - 7 - 8 1960 40109753 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1200000387.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnpla8

Panther Classification:

Molecular Function -

phospholipase

Gene Ontology Categories:

Function(s) Process(es)

prostaglandin biosynthetic process
lipid metabolic process
fatty acid metabolic process
metabolic process
cell death
lipid catabolic process
arachidonic acid metabolic process
phosphatidylcholine catabolic process
linoleic acid metabolic process
phosphatidylethanolamine catabolic process
arachidonic acid secretion
lysophospholipase activity
hydrolase activity
calcium-independent phospholipase A2 activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1200002736.mm.1 NM_026164.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1200002736.mm.1 NM_026164.2 Refseq
Affymetrix Mouse Clariom S Array PSR1200002736.mm.1 TC1200000387.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0