This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471148_m1
See other LIPF GE Assays ›
Gene Symbol:
Lipf
Gene Name:
lipase, gastric
Gene Aliases:
2310051B21Rik, AV082900
Chromosome Location:
Chr.19: 33919441 - 33976813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471148_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026334.3 NP_080610.1 3 - 4 268 113
GenBank mRNA AK009300.1 - 3 - 4 270 113
AK009359.1 - 3 - 4 270 113
AK009413.1 - 3 - 4 269 113
AK009428.1 - 3 - 4 272 113
AK009431.1 - 3 - 4 272 113
AK009437.1 - 3 - 4 270 113
AK009459.1 - 3 - 4 270 113
AK009473.1 - 3 - 4 269 113
AK009474.1 - 3 - 4 270 113
AK009479.1 - 3 - 4 267 113
AK009523.1 - 3 - 4 270 113
AK009525.1 - 3 - 4 270 113
AK009537.1 - 3 - 4 270 113
AK009546.1 - 3 - 4 273 113
AK009560.1 - 3 - 4 270 113
AK009571.1 - 3 - 4 269 113
AK009573.1 - 3 - 4 273 113
AK009729.1 - 3 - 4 270 113
AK009773.1 - 3 - 4 270 113
AK009875.1 - 3 - 4 270 113
AK009923.1 - 3 - 4 332 113
AK009990.1 - 3 - 4 270 113
AK010019.1 - 3 - 4 271 113
AK010026.1 - 3 - 4 270 113
AK010035.1 - 3 - 4 270 113
AK010036.1 - 3 - 4 268 113
AK010058.1 - 3 - 4 269 113
AK010061.1 - 3 - 4 270 113
AK010091.1 - 3 - 4 269 113
AK010103.1 - 3 - 4 271 113
AK010106.1 - 3 - 4 270 113
AK010110.1 - 3 - 4 273 113
AK010116.1 - 3 - 4 268 113
AK010123.1 - 3 - 4 271 113
AK010124.1 - 2 - 3 264 113
AK010125.1 - 3 - 4 270 113
AK010139.1 - 3 - 4 270 113
AK010148.1 - 3 - 4 270 113
AK010173.1 - 3 - 4 270 113
AK010203.1 - 3 - 4 271 113
AK010231.1 - 3 - 4 269 113
AK010236.1 - 3 - 4 271 113
AK075913.1 - 3 - 4 271 113
BC061067.1 - 3 - 4 268 30301084 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipf

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
oxidation-reduction process
triglyceride lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zjghb:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0