This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471150_m1
See other LIPF GE Assays ›
Gene Symbol:
Lipf
Gene Name:
lipase, gastric
Gene Aliases:
2310051B21Rik, AV082900
Chromosome Location:
Chr.19: 33919441 - 33976813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471150_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026334.3 NP_080610.1 5 - 6 580 112
GenBank mRNA AK009300.1 - 5 - 6 582 112
AK009359.1 - 5 - 6 582 112
AK009413.1 - 5 - 6 581 112
AK009428.1 - 5 - 6 584 112
AK009431.1 - 5 - 6 584 112
AK009437.1 - 5 - 6 582 112
AK009459.1 - 5 - 6 582 112
AK009473.1 - 5 - 6 581 112
AK009474.1 - 5 - 6 582 112
AK009479.1 - 5 - 6 579 112
AK009523.1 - 5 - 6 582 112
AK009525.1 - 5 - 6 582 112
AK009544.1 - 5 - 6 577 112
AK009546.1 - 5 - 6 585 112
AK009560.1 - 5 - 6 582 112
AK009571.1 - 5 - 6 581 112
AK009573.1 - 5 - 6 585 112
AK009729.1 - 5 - 6 582 112
AK009773.1 - 5 - 6 582 112
AK009875.1 - 5 - 6 582 112
AK009923.1 - 5 - 6 643 111
AK009990.1 - 5 - 6 582 112
AK010019.1 - 5 - 6 583 112
AK010026.1 - 5 - 6 582 112
AK010035.1 - 5 - 6 582 112
AK010036.1 - 5 - 6 580 112
AK010058.1 - 5 - 6 581 112
AK010061.1 - 5 - 6 582 112
AK010091.1 - 5 - 6 580 112
AK010093.1 - 5 - 6 582 112
AK010103.1 - 5 - 6 583 112
AK010116.1 - 5 - 6 580 112
AK010123.1 - 5 - 6 583 112
AK010124.1 - 4 - 5 576 112
AK010125.1 - 5 - 6 582 112
AK010139.1 - 5 - 6 582 112
AK010203.1 - 5 - 6 583 112
AK010231.1 - 5 - 6 581 112
AK010236.1 - 5 - 6 583 112
BC061067.1 - 5 - 6 580 30301084 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000488.mm.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipf

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
oxidation-reduction process
triglyceride lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004353.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004353.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004353.mm.1 TC1900000488.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004354.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004354.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004354.mm.1 TC1900000488.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0