This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471153_g1
See other LIPF GE Assays ›
Gene Symbol:
Lipf
Gene Name:
lipase, gastric
Gene Aliases:
2310051B21Rik, AV082900
Chromosome Location:
Chr.19: 33919441 - 33976813 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471153_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Lipf
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026334.3 NP_080610.1 8 - 9 933 72
GenBank mRNA AK009300.1 - 8 - 9 935 72
AK009359.1 - 8 - 9 935 72
AK009413.1 - 8 - 9 934 72
AK009428.1 - 8 - 9 937 72
AK009431.1 - 8 - 9 937 72
AK009437.1 - 8 - 9 935 72
AK009459.1 - 8 - 9 935 72
AK009473.1 - 8 - 9 934 72
AK009474.1 - 8 - 9 935 72
AK009479.1 - 8 - 9 932 72
AK009523.1 - 8 - 9 935 72
AK009525.1 - 8 - 9 935 72
AK009537.1 - 8 - 9 935 72
AK009544.1 - 8 - 9 930 72
AK009546.1 - 8 - 9 938 72
AK009560.1 - 8 - 9 935 72
AK009571.1 - 8 - 9 934 72
AK009573.1 - 8 - 9 938 72
AK009729.1 - 8 - 9 935 72
AK009773.1 - 8 - 9 935 72
AK009875.1 - 8 - 9 935 72
AK009923.1 - 8 - 9 996 72
AK009990.1 - 8 - 9 935 72
AK010019.1 - 8 - 9 936 72
AK010026.1 - 8 - 9 935 72
AK010035.1 - 8 - 9 935 72
AK010036.1 - 8 - 9 934 72
AK010058.1 - 8 - 9 934 72
AK010061.1 - 8 - 9 935 72
AK010091.1 - 8 - 9 933 72
AK010093.1 - 8 - 9 935 72
AK010103.1 - 8 - 9 936 72
AK010106.1 - 8 - 9 935 72
AK010110.1 - 8 - 9 938 72
AK010116.1 - 8 - 9 933 72
AK010123.1 - 8 - 9 935 72
AK010124.1 - 7 - 8 929 72
AK010125.1 - 8 - 9 935 72
AK010139.1 - 8 - 9 935 72
AK010148.1 - 8 - 9 935 72
AK010173.1 - 8 - 9 935 72
AK010203.1 - 8 - 9 936 72
AK010231.1 - 8 - 9 934 72
AK010236.1 - 8 - 9 936 72
AK075910.1 - 1 - 2 49 72
AK075913.1 - 8 - 9 935 72
BC061067.1 - 8 - 9 933 30301084 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000488.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Lipf

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
lipid catabolic process
oxidation-reduction process
triglyceride lipase activity
hydrolase activity
hydrolase activity, acting on ester bonds

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900004358.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004358.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004358.mm.1 TC1900000488.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900004357.mm.1 NM_026334.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900004357.mm.1 NM_026334.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900004357.mm.1 TC1900000488.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-6dxlp:80/100.66.78.215:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0