This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471201_m1
See other NUDT13 GE Assays ›
Gene Symbol:
Nudt13
Gene Name:
nudix (nucleoside diphosphate linked moiety X)-type motif 13
Gene Aliases:
4933433B15Rik
Chromosome Location:
Chr.14: 20294679 - 20317584 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471201_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nudt13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026341.2 NP_080617.2 2 - 3 192 71
GenBank mRNA AK014204.1 - 3 - 4 415 71
AK017031.1 - 2 - 3 161 71
AK038639.1 - 1 - 2 174 71
BC021366.1 - 3 - 4 326 71
BC037091.1 - 1 - 2 115 71
BC038023.1 - 3 - 4 274 71
BC040807.1 - 2 - 3 139 71
BC049948.1 - 2 - 3 146 3666660 71
BC056982.1 - 3 - 4 321 71
BF323208.1 - 2 - 3 171 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1400000163.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nudt13

Gene Ontology Categories:

Function(s) Process(es)

biological_process
GPI-anchor transamidase activity
hydrolase activity
metal ion binding
coenzyme F390-A hydrolase activity
coenzyme F390-G hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1400001254.mm.1 NM_026341.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001254.mm.1 NM_026341.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001254.mm.1 TC1400000163.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001246.mm.1 NM_026341.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001246.mm.1 NM_026341.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001246.mm.1 TC1400000163.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001253.mm.1 NM_026341.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001253.mm.1 NM_026341.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001253.mm.1 TC1400000163.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001247.mm.1 NM_026341.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001247.mm.1 NM_026341.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001247.mm.1 TC1400000163.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1400001252.mm.1 NM_026341.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1400001252.mm.1 NM_026341.2 Refseq
Affymetrix Mouse Clariom S Array PSR1400001252.mm.1 TC1400000163.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0