This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471676_g1
See other NDUFA5 GE Assays ›
Gene Symbol:
Ndufa5
Gene Name:
NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 5
Gene Aliases:
2900002J19Rik
Chromosome Location:
Chr.6: 24518662 - 24527687 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471676_g1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Ndufa5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026614.2 NP_080890.1 1 - 2 167 72
GenBank mRNA AK008155.1 - 1 - 2 38 72
AK008156.1 - 1 - 2 33 72
AK008286.1 - 1 - 2 33 72
AK008414.1 - 1 - 2 33 72
AK009338.1 - 1 - 2 51 72
AK009396.1 - 1 - 2 37 72
AK009461.1 - 1 - 2 37 72
AK009493.1 - 1 - 2 36 72
AK009608.1 - 1 - 2 37 72
AK009642.1 - 1 - 2 52 72
AK009652.1 - 1 - 2 37 72
AK009775.1 - 1 - 2 61 72
AK009840.1 - 1 - 2 51 72
AK009985.1 - 1 - 2 51 72
AK010024.1 - 1 - 2 36 72
AK010090.1 - 1 - 2 39 72
AK010092.1 - 1 - 2 53 72
AK019084.1 - 1 - 2 38 72
AK019201.1 - 1 - 2 38 72
AK019215.1 - 1 - 2 37 72
AK019242.1 - 1 - 2 35 72
AK019245.1 - 1 - 2 58 72
AK019291.1 - 1 - 2 38 72
AK019301.1 - 1 - 2 168 72
AK019324.1 - 1 - 2 147 72
AK019354.1 - 1 - 2 40 72
AK019359.1 - 1 - 2 59 72
AK019364.1 - 1 - 2 53 72
AK019368.1 - 1 - 2 51 72
AK019430.1 - 1 - 2 72
BC028633.1 - 1 - 2 35 1247576 72

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK008156.1, AK008286.1, AK008414.1, AK009338.1, AK009396.1, AK009461.1, AK009493.1, AK009608.1, AK009642.1, AK009652.1, AK009775.1, AK009840.1, AK009985.1, AK010024.1, AK010090.1, AK010092.1, AK019084.1, AK019201.1, AK019215.1, AK019242.1, AK019245.1, AK019291.1, AK019301.1, AK019324.1, AK019354.1, AK019359.1, AK019364.1, AK019368.1, AK019430.1, BC028633.1, NM_026614.2 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Ndufa5

Panther Classification:

Molecular Function -

oxidoreductase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

transport
respiratory electron transport chain
oxidation-reduction process
NADH dehydrogenase (ubiquinone) activity
oxidoreductase activity, acting on NAD(P)H

MicroArray Probes:

ABI

Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 912244 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0