This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00471842_m1
See other PNLIP GE Assays ›
Gene Symbol:
Pnlip
Gene Name:
pancreatic lipase
Gene Aliases:
1810007A24Rik, PL, PTL
Chromosome Location:
Chr.19: 58670365 - 58681788 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00471842_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pnlip
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_026925.3 NP_081201.2 2 - 3 68 79
GenBank mRNA AK007357.1 - 2 - 3 69 79
BC061061.1 - 2 - 3 68 30316303 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1900000838.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK007357.1,BC061061.1,NM_026925.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pnlip

Panther Classification:

Molecular Function -

lipase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
post-embryonic development
lipid catabolic process
intestinal cholesterol absorption
positive regulation of triglyceride lipase activity
triglyceride lipase activity
lipase activity
hydrolase activity
metal ion binding
carboxylic ester hydrolase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1900007277.mm.1 NM_026925.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007277.mm.1 NM_026925.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007277.mm.1 TC1900000838.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007278.mm.1 NM_026925.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007278.mm.1 NM_026925.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007278.mm.1 TC1900000838.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1900007276.mm.1 NM_026925.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1900007276.mm.1 NM_026925.3 Refseq
Affymetrix Mouse Clariom S Array PSR1900007276.mm.1 TC1900000838.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8t6tc:80/100.66.75.151:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0