This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00473998_m1
See other AGPAT3 GE Assays ›
Gene Symbol:
Agpat3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
AW061257, AW493985, D10Jhu12e, lpaat3
Chromosome Location:
Chr.10: 78269174 - 78352484 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00473998_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053014.3 NP_443747.2 3 - 4 368 89
GenBank mRNA AB377215.1 - 1 - 2 184 89
AK015906.1 - 2 - 3 296 89
AK030607.1 - 1 - 2 160 89
AK075715.1 - 3 - 4 368 89
AK076414.1 - 2 - 3 337 89
AK159110.1 - 2 - 3 608 89
AY167588.1 - 1 - 2 206 89
BC033444.1 - 1 - 2 46 89
BC052382.1 - 2 - 3 259 5703571 89
BC058519.1 - 2 - 3 258 5703571 89

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002448.mm.1

Additional Information:

This assay is in the 5'UTR region of the AK030607.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agpat3

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
metabolic process
phospholipid biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
lysophosphatidic acid acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000017849.mm.1 NM_053014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017849.mm.1 NM_053014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017849.mm.1 TC1000002448.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0