This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00473999_m1
See other AGPAT3 GE Assays ›
Gene Symbol:
Agpat3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
AW061257, AW493985, D10Jhu12e, lpaat3
Chromosome Location:
Chr.10: 78269174 - 78352484 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00473999_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053014.3 NP_443747.2 4 - 5 534 63
GenBank mRNA AB377215.1 - 2 - 3 350 63
AK015906.1 - 3 - 4 462 63
AK030607.1 - 2 - 3 326 63
AK075715.1 - 4 - 5 534 63
AK076414.1 - 3 - 4 503 63
AK159110.1 - 3 - 4 774 63
AY167588.1 - 2 - 3 372 63
BC033444.1 - 2 - 3 212 63
BC052382.1 - 3 - 4 425 5703571 63
BC058519.1 - 3 - 4 424 5703571 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1000002448.mm.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agpat3

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
metabolic process
phospholipid biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
lysophosphatidic acid acyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1000017847.mm.1 NM_053014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017847.mm.1 NM_053014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017847.mm.1 TC1000002448.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000017846.mm.1 NM_053014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017846.mm.1 NM_053014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017846.mm.1 TC1000002448.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000017848.mm.1 NM_053014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017848.mm.1 NM_053014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017848.mm.1 TC1000002448.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1000017845.mm.1 NM_053014.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR1000017845.mm.1 NM_053014.3 Refseq
Affymetrix Mouse Clariom S Array PSR1000017845.mm.1 TC1000002448.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0