This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00474000_m1
See other AGPAT3 GE Assays ›
Gene Symbol:
Agpat3
Gene Name:
1-acylglycerol-3-phosphate O-acyltransferase 3
Gene Aliases:
AW061257, AW493985, D10Jhu12e, lpaat3
Chromosome Location:
Chr.10: 78269174 - 78352484 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00474000_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Agpat3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053014.3 NP_443747.2 5 - 6 689 63
GenBank mRNA AB377215.1 - 3 - 4 505 63
AK008965.1 - 5 - 6 845 63
AK015906.1 - 4 - 5 617 63
AK030607.1 - 3 - 4 481 63
AK075715.1 - 5 - 6 689 63
AK076414.1 - 4 - 5 658 63
AK159110.1 - 4 - 5 929 63
AY167588.1 - 3 - 4 527 63
BC033444.1 - 3 - 4 367 63
BC052382.1 - 4 - 5 580 5703571 63
BC058519.1 - 4 - 5 579 5703571 63

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Agpat3

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

lipid metabolic process
metabolic process
phospholipid biosynthetic process
1-acylglycerol-3-phosphate O-acyltransferase activity
transferase activity
transferase activity, transferring acyl groups
lysophosphatidic acid acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nbrrd:80/100.66.77.69:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0