This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00474147_g1
See other SMARCA1 GE Assays ›
Gene Symbol:
Smarca1
Gene Name:
SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily a, member 1
Gene Aliases:
5730494M04Rik, Snf2l
Chromosome Location:
Chr.X: 47809370 - 47892931 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00474147_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Smarca1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001290708.1 NP_001277637.1 3 - 4 598 90
NM_053123.4 NP_444353.3 3 - 4 598 90
GenBank mRNA AF325920.1 - 3 - 4 605 90
AK030741.1 - 3 - 4 565 90
AK032978.1 - 3 - 4 566 90
AK035068.1 - 3 - 4 463 90
AK052092.1 - 3 - 4 554 90
BC016071.1 - 3 - 4 575 90
BC057115.1 - 3 - 4 598 6406330 90

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Smarca1

Panther Classification:

Molecular Function -

DNA helicase

Gene Ontology Categories:

Function(s) Process(es)

DNA strand renaturation
chromatin remodeling
transcription, DNA-templated
regulation of transcription, DNA-templated
brain development
chromatin modification
neuron differentiation
ATP-dependent chromatin remodeling
positive regulation of transcription, DNA-templated
regulation of neural precursor cell proliferation
nucleotide binding
nucleic acid binding
DNA binding
chromatin binding
helicase activity
ATP binding
DNA-dependent ATPase activity
hydrolase activity
hydrolase activity, acting on acid anhydrides
hydrolase activity, acting on acid anhydrides, in phosphorus-containing anhydrides
ATPase activity
nucleosome binding
annealing helicase activity
nucleosome-dependent ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0