This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00474799_m1
See other CADM3 GE Assays ›
Gene Symbol:
Cadm3
Gene Name:
cell adhesion molecule 3
Gene Aliases:
BIgR, Igsf4b, Necl-1, Necl1, SynCAM3, Tsll1
Chromosome Location:
Chr.1: 173334254 - 173367695 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00474799_m1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Cadm3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_053199.3 NP_444429.1 1 - 2 282 143
GenBank mRNA AF195662.1 - 1 - 2 226 143
AK038917.1 - 1 - 2 239 143
AK053077.1 - 1 - 2 236 143
BC115417.2 - 1 - 2 187 40093410 143
BC115418.2 - 1 - 2 187 40093417 143

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC0100003587.mm.1

Additional Information:

This assay is in the 5'UTR region of the AF195662.1,AK038917.1,AK053077.1,BC115417.2,BC115418.2,NM_053199.3 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus 1700 Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Cadm3

Gene Ontology Categories:

Function(s) Process(es)

cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
heterophilic cell-cell adhesion via plasma membrane cell adhesion molecules
protein localization
protein binding
protein homodimerization activity

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR0100029433.mm.1 NM_053199.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029433.mm.1 NM_053199.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100029433.mm.1 TC0100003587.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029431.mm.1 NM_053199.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029431.mm.1 NM_053199.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100029431.mm.1 TC0100003587.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029436.mm.1 NM_053199.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029436.mm.1 NM_053199.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100029436.mm.1 TC0100003587.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR0100029434.mm.1 NM_053199.3.1 Refseq
Affymetrix Mouse Clariom S Array PSR0100029434.mm.1 NM_053199.3 Refseq
Affymetrix Mouse Clariom S Array PSR0100029434.mm.1 TC0100003587.mm.1 MTA TC
Array Name Probe IDs Targets Target Type
ABI Mouse Genome Survey Microarray 772789 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0