Assay Details
| Gene Symbol: Elmo2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_080287.2 | NP_525026.2 | 7 - 8 | 621 | 59 | |
| NM_207706.1 | NP_997589.1 | 7 - 8 | 621 | 59 | ||
| GenBank mRNA | AF398884.1 | - | 6 - 7 | 520 | 59 | |
| AK032033.1 | - | 7 - 8 | 594 | 59 | ||
| AK033918.1 | - | 7 - 8 | 815 | 59 | ||
| AK038097.1 | - | 7 - 8 | 612 | 59 | ||
| AK038455.1 | - | 7 - 8 | 727 | 59 | ||
| AK045428.1 | - | 7 - 8 | 622 | 59 | ||
| AK047040.1 | - | 7 - 8 | 583 | 59 | ||
| AK050823.1 | - | 7 - 8 | 584 | 59 | ||
| AK053574.1 | - | 7 - 8 | 585 | 59 | ||
| AK129023.1 | - | 7 - 8 | 621 | 59 | ||
| BC023954.1 | - | 7 - 8 | 606 | 5320429 | 59 | |
| BC033418.1 | - | 6 - 7 | 551 | 59 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Elmo2
Genomic Map