| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR1200010733.mm.1 |
NM_001081977.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010733.mm.1 |
TC1200001549.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010735.mm.1 |
NM_001081977.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010735.mm.1 |
NM_080563.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010735.mm.1 |
TC1200001549.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR1200010734.mm.1 |
NM_001081977.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010734.mm.1 |
NM_080563.4 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR1200010734.mm.1 |
TC1200001549.mm.1 |
MTA TC |