This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00475697_g1
See other ACP5 GE Assays ›
Gene Symbol:
Acp5
Gene Name:
acid phosphatase 5, tartrate resistant
Gene Aliases:
TRACP, TRAP
Chromosome Location:
Chr.9: 22126727 - 22135746 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00475697_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Acp5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001102404.1 NP_001095874.1 2 - 3 389 71
NM_001102405.1 NP_001095875.1 2 - 3 394 71
NM_007388.3 NP_031414.1 2 - 3 336 71
GenBank mRNA AK008391.1 - 1 - 2 116 71
AK010865.1 - 2 - 3 360 71
AK011915.1 - 2 - 3 360 71
BC012911.1 - 2 - 3 288 3588062 71
BC019160.1 - 2 - 3 352 3596147 71
BC029644.1 - 2 - 3 288 3584530 71
BG076865.2 - 3 - 4 441 71

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Acp5

Panther Classification:

Molecular Function -

hydrolase phosphatase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

dephosphorylation
response to lipopolysaccharide
negative regulation of interleukin-1 beta production
negative regulation of interleukin-12 production
negative regulation of tumor necrosis factor production
negative regulation of superoxide anion generation
response to cytokine
negative regulation of nitric oxide biosynthetic process
bone resorption
negative regulation of inflammatory response
defense response to Gram-positive bacterium
bone morphogenesis
acid phosphatase activity
hydrolase activity
metal ion binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0