This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00476558_m1
See other NTHL1 GE Assays ›
Gene Symbol:
Nthl1
Gene Name:
nth (endonuclease III)-like 1 (E.coli)
Gene Aliases:
Nth1, Octs3
Chromosome Location:
Chr.17: 24632682 - 24638838 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00476558_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Nthl1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_008743.2 NP_032769.2 3 - 4 582 72
GenBank mRNA AB006812.1 - 3 - 4 528 72
AK033701.1 - 3 - 4 1590 72
AV377313.2 - 1 - 2 170 72
BC138852.1 - 3 - 4 545 8861874 72
BC138855.1 - 3 - 4 545 8861877 72
BC145443.1 - 3 - 4 454 72
CX565655.1 - 3 - 4 561 72
Y09688.1 - 3 - 4 554 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1700000346.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Nthl1

Panther Classification:

Molecular Function -

DNA metabolism protein DNA glycosylase nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

positive regulation of defense response to virus by host
DNA repair
base-excision repair
base-excision repair, AP site formation
nucleotide-excision repair, DNA incision, 5'-to lesion
cellular response to DNA damage stimulus
metabolic process
mitophagy in response to mitochondrial depolarization
purine-specific mismatch base pair DNA N-glycosylase activity
DNA binding
double-stranded DNA binding
catalytic activity
DNA-(apurinic or apyrimidinic site) lyase activity
uracil DNA N-glycosylase activity
hydrolase activity
hydrolase activity, acting on glycosyl bonds
lyase activity
single-strand selective uracil DNA N-glycosylase activity
DNA N-glycosylase activity
DNA-3-methylbase glycosylase activity
metal ion binding
iron-sulfur cluster binding
4 iron, 4 sulfur cluster binding
nicotinamide riboside hydrolase activity
nicotinic acid riboside hydrolase activity
deoxyribonucleoside 5'-monophosphate N-glycosidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1700002638.mm.1 NM_008743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700002638.mm.1 NM_008743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700002638.mm.1 TC1700000346.mm.1 MTA TC
Affymetrix Mouse Clariom S Array PSR1700002637.mm.1 NM_008743.2.1 Refseq
Affymetrix Mouse Clariom S Array PSR1700002637.mm.1 NM_008743.2 Refseq
Affymetrix Mouse Clariom S Array PSR1700002637.mm.1 TC1700000346.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline