This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Mm00476644_g1
See other PDE1B GE Assays ›
Gene Symbol:
Pde1b
Gene Name:
phosphodiesterase 1B, Ca2+-calmodulin dependent
Gene Aliases:
Pde1b1
Chromosome Location:
Chr.15: 103503034 - 103530056 on Build GRCm38
Species:
Mouse
Species Specific ID (Flybase ID):
-
Assay ID Mm00476644_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: Pde1b
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001285890.1 NP_001272819.1 10 - 11 1520 82
NM_008800.2 NP_032826.1 11 - 12 1537 82
GenBank mRNA AK004772.1 - 10 - 11 1127 82
BC058531.1 - 11 - 12 1537 6826674 82
L01695.1 - 11 - 12 1280 82

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as MTA transcript cluster(s) TC1500001133.mm.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene Pde1b

Panther Classification:

Molecular Function -

phosphodiesterase

Gene Ontology Categories:

Function(s) Process(es)

regulation of neurotransmitter levels
response to amphetamine
cAMP catabolic process
signal transduction
locomotory behavior
visual learning
cellular response to macrophage colony-stimulating factor stimulus
regulation of dopamine metabolic process
serotonin metabolic process
cGMP catabolic process
cellular response to granulocyte macrophage colony-stimulating factor stimulus
cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-nucleotide phosphodiesterase activity
3',5'-cyclic-AMP phosphodiesterase activity
calmodulin-dependent cyclic-nucleotide phosphodiesterase activity
calmodulin binding
phosphoric diester hydrolase activity
hydrolase activity
metal ion binding
calcium- and calmodulin-regulated 3',5'-cyclic-GMP phosphodiesterase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Mouse Clariom S Array PSR1500009141.mm.1 NM_001285890.1 Refseq
Affymetrix Mouse Clariom S Array PSR1500009141.mm.1 NM_008800.2 Refseq
Affymetrix Mouse Clariom S Array PSR1500009141.mm.1 TC1500001133.mm.1 MTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline